Invitrogen
Manufacturer Code:PA524380
Catalog # PIPA524380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 279-308 amino acids from the C-terminal region of human OR9Q1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GA_x6K02T2RE5P-4160554-4159619; olfactory receptor 1500; Olfactory receptor 9Q1; olfactory receptor MOR212-4P; olfactory receptor, family 9, subfamily Q, member 1
Gene Aliases: MOR212-4P; OR9Q1
UniProt ID: (Human) Q8NGQ5
Entrez Gene ID: (Human) 219956, (Mouse) 258097
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.