Invitrogen
Manufacturer Code:OSR00030G
Catalog # PIOSR00030G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from n-terminal region of mouse olfactory receptor family 256 subfamily 17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GA_x54KRFPKG5P-348087-349025; Odorant receptor OR3; Olfactory receptor 15; Olfactory receptor 256-17; olfactory receptor MOR256-17; Olfactory receptor Olfr15 Odorant receptor OR3. GENE NAME: Olfr15.
Gene Aliases: MOR256-17; Olfr15; OR3
UniProt ID: (Mouse) P23275
Entrez Gene ID: (Mouse) 18312
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.