Novus Biologicals
Manufacturer Code:NBP131163
Catalog # NBP131163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Noelin. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMYolfactomedin related ER localized protein neuroblastoma protein Neuronal olfactomedin-related ER localized protein NOE1noelin NOEL1 NOELIN NOELIN1 OlfA olfactomedin 1 olfactomedin-1 pancortin 1; neuroblastoma protein; Neuronal olfactomedin-related ER localized protein; Noelin; olfactomedin related ER localized protein; Olfactomedin-1; pancortin 1
Gene Aliases: AMY; NOE1; NOEL1; NOELIN1; OlfA; OLFM1
UniProt ID: (Human) Q99784
Entrez Gene ID: (Human) 10439
Molecular Function: receptor structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.