R&D Systems
Manufacturer Code:BAF4636
Catalog # BAF4636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Olfactomedin-1/Noelin-1 Ser2-Arg481 Accession # NP_062371 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: AMY AMYolfactomedin related ER localized protein neuroblastoma protein Neuronal olfactomedin-related ER localized protein NOE1 NOE1noelin NOEL1 NOELIN Noelin-1 NOELIN1 OlfA olfactomedin 1 olfactomedin-1 Olfactomedin1 OLFM1 pancortin 1; neuroblastoma protein; Neuronal olfactomedin-related ER localized protein; Noelin; olfactomedin related ER localized protein; Olfactomedin-1; pancortin 1
Gene Aliases: AMY; NOE1; NOEL1; NOELIN1; OlfA; OLFM1
UniProt ID: (Human) Q99784
Entrez Gene ID: (Human) 10439
Molecular Function: receptor structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.