Invitrogen
Manufacturer Code:PA583946
Catalog # PIPA583946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human POU2F2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homeobox protein; Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; Oct-2; OCT2 OTF2; Octamer-binding protein 2; Octamer-binding transcription factor 2; OTF-2; POU domain, class 2, transcription factor 2
Gene Aliases: Oct-2; OCT2; OTF2; POU2F2
UniProt ID: (Human) P09086
Entrez Gene ID: (Human) 5452
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.