Proteintech
Manufacturer Code:272601AP150UL
Catalog # 50-173-5622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Occludin Fusion Protein Ag26173 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Occludin; Occludin OCLN; phosphatase 1, regulatory subunit 115; tight junction protein occludin
Gene Aliases: AI503564; BLCPMG; Ocl; OCLN; PPP1R115
UniProt ID: (Human) Q16625, (Mouse) Q61146
Entrez Gene ID: (Human) 100506658, (Mouse) 18260
Molecular Function:
cell junction protein
tight junction
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.