Novus Biologicals
Manufacturer Code:NBP147905
Catalog # NBP147905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F3 |
Immunogen | Full-length protein expressed in 293T cell transfected with human OXSR1 expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.1 KIAA1101OSR1serine/threonine-protein kinase OSR1 Oxidative stress-responsive 1 protein oxidative-stress responsive 1; Oxidative stress-responsive 1 protein; oxidative-stress responsive 1; OXSR1; Serine/threonine-protein kinase OSR1
Gene Aliases: KIAA1101; OSR1; OXSR1
UniProt ID: (Human) O95747
Entrez Gene ID: (Human) 9943
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.