Invitrogen
Manufacturer Code:PA532133
Catalog # PIPA532133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 236 of Human OXSM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-ketoacyl-acyl carrier protein synthase; 3-oxoacyl-[acyl-carrier-protein] synthase, mitochondrial; 3-oxoacyl-acyl-carrier-protein synthase; acyl-malonyl acyl carrier protein-condensing enzyme; beta-ketoacyl synthase; Beta-ketoacyl-ACP synthase; FASN2D KASI KS 3-ketoacyl-acyl carrier protein synthase 3-oxoacyl-&lsqbacyl-carrier-protein] synthase mitochondrial 3-oxoacyl-acyl-carrier-protein synthase acyl-malonyl acyl carrier protein-condensing enzyme beta-ketoacyl synthase beta-ketoacyl-ACP synthase type II mitocho; type II mitochondrial beta-ketoacyl synthase
Gene Aliases: 4933425A18Rik; C80494; FASN2D; KASI; KS; OXSM
UniProt ID: (Human) Q9NWU1, (Mouse) Q9D404
Entrez Gene ID: (Human) 54995, (Mouse) 71147
Molecular Function:
acyltransferase
dehydrogenase
esterase
hydrolase
ligase
methyltransferase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.