Novus Biologicals
Manufacturer Code:NBP273175AF350
Catalog # NP27317A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D1 |
Immunogen | Full length recombinant protein of human OXNAD1 (NP612390)produced in HEK293 cell. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: MGC15763 oxidoreductase NAD-binding domain containing 1 oxidoreductase NAD-binding domain-containing protein 1; oxidoreductase NAD-binding domain containing 1; Oxidoreductase NAD-binding domain-containing protein 1
Gene Aliases: OXNAD1
UniProt ID: (Human) Q96HP4
Entrez Gene ID: (Human) 92106
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.