Novus Biologicals
Manufacturer Code:NBP297990JF646
Catalog # NB050579
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human OXER1/5-oxo-ETE GPCR fragment. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-oxo-ETE acid G-protein-coupled receptor 1; 5-oxo-ETE acid G-protein-coupled receptor 15-oxo-ETE G-protein coupled receptor GPCR GPR170G-protein coupled receptor R527 G-protein coupled receptor 170 oxoeicosanoid (OXE) receptor 1 oxoeicosanoid receptor 1 TG1019G-protein coupled receptor TG1019; 5-oxo-ETE G-protein coupled receptor; G-protein coupled receptor 170; G-protein coupled receptor R527; G-protein coupled receptor TG1019; OXE receptor; oxoeicosanoid (OXE) receptor 1; Oxoeicosanoid receptor 1
Gene Aliases: GPCR; GPR170; OXER1; TG1019
UniProt ID: (Human) Q8TDS5
Entrez Gene ID: (Human) 165140
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.