Invitrogen
Manufacturer Code:PA5116057
Catalog # PIPA5116057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TNFSF4 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ath1 Ath-1 atherosclerosis 1 CD134 ligand CD134L CD252 Glycoprotein Gp34 GP34 OX40 antigen ligand OX40 ligand OX40L OX-40L OX4OL TAX transcriptionally-activated glycoprotein 1 tax-transcriptionally activated glycoprotein 1 (34kD) tax-transcriptionally activated glycoprotein 1 ligand Tnfsf4 TNLG2B tumor necrosis factor (ligand) superfamily member 4 tumor necrosis factor (ligand) superfamily member 4 tumor necrosis factor ligand 2B tumor necrosis factor ligand superfamily member 4 tumor necrosis factor superfamily member 4 Txgp1 Txgp1l; atherosclerosis 1; CD134 ligand; CD252; Glycoprotein Gp34; OX40 antigen ligand; OX40 ligand; OX40L; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 ligand; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor ligand 2B; Tumor necrosis factor ligand superfamily member 4
Gene Aliases: Ath-1; Ath1; CD134L; CD252; GP34; OX-40L; Ox40l; OX4OL; TNFSF4; TNLG2B; TXGP1; Txgp1l
UniProt ID: (Human) P23510, (Mouse) P43488
Entrez Gene ID: (Human) 7292, (Mouse) 22164
Molecular Function:
cytokine
signaling molecule
tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.