R&D Systems
Manufacturer Code:AF1054
Catalog # AF1054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human OX40 Ligand/TNFSF4 Gln51-Leu183 Accession # Q6FGS4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD134 ligand; CD134 ligand CD134L CD252 CD252 antigen Glycoprotein Gp34 gp34 OX40 antigen ligand OX40 ligand OX40L OX-40L OX4OL TAX transcriptionally-activated glycoprotein 1 tax-transcriptionally activated glycoprotein 1 (34kD) tax-transcriptionally activated glycoprotein 1 34kD TNFSF4 tumor necrosis factor (ligand) superfamily member 4 tumor necrosis factor ligand superfamily member 4 TXGP1; CD252; Glycoprotein Gp34; OX40 antigen ligand; OX40 ligand; OX40L; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor ligand 2B; Tumor necrosis factor ligand superfamily member 4
Gene Aliases: CD134L; CD252; GP34; OX-40L; OX4OL; TNFSF4; TNLG2B; TXGP1
UniProt ID: (Human) P23510
Entrez Gene ID: (Human) 7292
Molecular Function: cytokine signaling molecule tumor necrosis factor family member
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.