Invitrogen
Manufacturer Code:PA518952
Catalog # PIPA518952
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (YNFLSGKFREQFK) corresponding to the internal amino acids of HCRTR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: HCRTR1 Hcrtr2 OX1R OX2R HCRTR1 hypocretin orexin receptor 1 hypocretin receptor 1 hypocretin receptor-1 orexin receptor 1 orexin receptor-1 HCRTR2 hypocretin orexin receptor 2 hypocretin receptor 2 hypocretin receptor-2 orexin receptor 2 orexin receptor-2; hypocretin (orexin) receptor 1; hypocretin (orexin) receptor 2; hypocretin receptor 1; hypocretin receptor 2; Hypocretin receptor type 1; Hypocretin receptor type 2; hypocretin receptor-1; orexin receptor 1; Orexin receptor type 1; Orexin receptor type 2; orexin receptor-1; orexin type-2 receptor; Ox-1-R; Ox-2-R; ox1-R; ox1r; ox2-R; ox2r
Gene Aliases: HCRTR1; HCRTR2; Hctr1; mOX2aR; mOX2bR; Mox2r; mOXR2; ox-1-R; ox1-R; OX1R; OX2R
UniProt ID: (Human) O43613, (Human) O43614, (Mouse) P58307, (Rat) P56718, (Rat) P56719, (Mouse) P58308
Entrez Gene ID: (Human) 3061, (Human) 3062, (Mouse) 230777, (Rat) 25593, (Rat) 25605, (Mouse) 387285
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.