Novus Biologicals
Manufacturer Code:NBP188754
Catalog # NBP188754
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QYAYKQRRDKLYVCEDCGYTGPTQEDLYLHVNSAHPGSSFLKKTSKKLAALLQGKLTSAHQENTSLSEEEERK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: bA504H3.3 hOvo2 ovo-like 2 (Drosophila) transcription factor Ovo-like 2 Zinc finger protein 339EUROIMAGE566589 ZNF339; hOvo2; Transcription factor Ovo-like 2; Zinc finger protein 339
Gene Aliases: EUROIMAGE566589; OVOL2; PPCD1; ZNF339
UniProt ID: (Human) Q9BRP0
Entrez Gene ID: (Human) 58495
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.