Novus Biologicals
Manufacturer Code:NBP2822990.1ML
Catalog # NB030228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human OVOL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA504H3.3 hOvo2 ovo-like 2 (Drosophila) transcription factor Ovo-like 2 Zinc finger protein 339EUROIMAGE566589 ZNF339; hOvo2; Transcription factor Ovo-like 2; Zinc finger protein 339
Gene Aliases: EUROIMAGE566589; OVOL2; PPCD1; ZNF339
UniProt ID: (Human) Q9BRP0
Entrez Gene ID: (Human) 58495
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.