Invitrogen
Manufacturer Code:PA555495
Catalog # PIPA555495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human OTUD7B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cellular zinc finger anti-NF-kappa-B protein; cellular zinc finger anti-NF-kappaB Cezanne; Cezanne; OTU domain containing 7B; OTU domain-containing protein 7B; OTU domain-containing protein 7B Cellular zinc finger anti-NF-kappa-B protein Zinc finger A20 domain-containing protein 1 Zinc finger protein Cezanne OTUD7B ZA20D1; TRAF binding domain containing 2; Zinc finger A20 domain-containing protein 1; Zinc finger protein Cezanne; zinc finger, A20 domain containing 1
Gene Aliases: 2900060B22Rik; 4930463P07Rik; AI462125; CEZANNE; OTUD7B; RGD1308111; Trabid2; ZA20D1
UniProt ID: (Human) Q6GQQ9, (Mouse) B2RUR8
Entrez Gene ID: (Human) 56957, (Rat) 310677, (Mouse) 229603
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.