Invitrogen
Manufacturer Code:PA540934
Catalog # PIPA540934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ZA20D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cellular zinc finger anti-NF-kappa-B protein; cellular zinc finger anti-NF-kappaB Cezanne; Cezanne; OTU domain containing 7B; OTU domain-containing protein 7B; TRAF binding domain containing 2; Zinc finger A20 domain-containing protein 1; Zinc finger protein Cezanne; zinc finger, A20 domain containing 1
Gene Aliases: 2900060B22Rik; 4930463P07Rik; AI462125; CEZANNE; OTUD7B; RGD1308111; Trabid2; ZA20D1
UniProt ID: (Human) Q6GQQ9, (Mouse) B2RUR8
Entrez Gene ID: (Human) 56957, (Dog) 483178, (Bovine) 525579, (Pig) 100152569, (Horse) 100054396, (Rabbit) 100348660, (Rat) 310677, (Mouse) 229603, (Guinea pig) 100719181
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.