Invitrogen
Manufacturer Code:PA5110066
Catalog # PIPA5110066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human OTUD6A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DUBA-2; DUBA2 DUBA-2 EG408193 Hin6 Hin-6 protease HSHIN6 OTU domain-containing protein 6A Otud6a RGD1560855; HIN-6 protease; OTU deubiquitinase 6A; OTU domain containing 6A; OTU domain-containing protein 6A
Gene Aliases: DUBA2; EG408193; Hin6; HSHIN6; OTUD6A; RGD1560855
UniProt ID: (Human) Q7L8S5, (Mouse) Q6IE21
Entrez Gene ID: (Human) 139562, (Rat) 302426, (Mouse) 408193
Molecular Function:
hydrolase
nucleic acid binding
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.