Proteintech
Manufacturer Code:244861AP150UL
Catalog # 50-173-4382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | OTUD6A Fusion Protein Ag21639 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DUBA 2 DUBA2 HSHIN6 OTU domain containing 6A OTUD6A; DUBA-2; HIN-6 protease; OTU domain-containing protein 6A
Gene Aliases: DUBA2; HSHIN6; OTUD6A
UniProt ID: (Human) Q7L8S5
Entrez Gene ID: (Human) 139562
Molecular Function:
hydrolase
nucleic acid binding
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.