Invitrogen
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | 1:10-1:50 |
Western Blot (WB) | 1:1,000 |
Host / Isotype | Rabbit / IgG |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 360-388 amino acids from the Central region of human OTOP1. |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | Lot-Specific |
Storage conditions | -20° C, Avoid Freeze/Thaw Cycles |
RRID | AB_2633776 |
Predicted to react with rat based on sequence homology.
Required for normal formation of otoconia in the inner ear. Inhibits P2Y purinoceptors. Modulates calcium homeostasis and influx of calcium in response to extracellular ATP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.