Proteintech
Manufacturer Code:244561AP150UL
Catalog # 50-173-4369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | OTOP1 Fusion Protein Ag20011 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hOtop1; OTOP1 otopetrin 1; Otopetrin-1; Proton channel OTOP1; tilted
Gene Aliases: A530025J20Rik; OTOP1; Otp1; tlt
UniProt ID: (Human) Q7RTM1, (Mouse) Q80VM9
Entrez Gene ID: (Human) 133060, (Mouse) 21906
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.