Invitrogen
Manufacturer Code:PA540510
Catalog # PIPA540510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human OSR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: odd-skipped homolog; odd-skipped related 1; odd-skipped related 1 (Drosophila) ODD; odd-skipped related transciption factor 1; Protein odd-skipped-related 1
Gene Aliases: ODD; ODD1; Osr; OSR1
UniProt ID: (Human) Q8TAX0, (Bovine) Q08DS3, (Mouse) Q9WVG7, (Rat) B0K011
Entrez Gene ID: (Human) 130497, (Dog) 482984, (Pig) 100520131, (Horse) 100056547, (Rabbit) 100357466, (Bovine) 539430, (Mouse) 23967, (Guinea pig) 100731944, (Rat) 298878
Molecular Function:
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.