Invitrogen
Manufacturer Code:PA5116814
Catalog # PIPA5116814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human OSR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ODD Odd1 odd-skipped homolog odd-skipped related 1 odd-skipped related 1 (Drosophila) odd-skipped related transciption factor 1 Osr OSR1 Protein odd-skipped-related 1; odd-skipped homolog; odd-skipped related 1; Protein odd-skipped-related 1
Gene Aliases: ODD; Odd1; Osr; OSR1
UniProt ID: (Human) Q8TAX0, (Mouse) Q9WVG7, (Rat) B0K011
Entrez Gene ID: (Human) 130497, (Mouse) 23967, (Rat) 298878
Molecular Function:
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.