Novus Biologicals
Manufacturer Code:NBP131198
Catalog # NBP131198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human OSR1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: odd-skipped homolog; odd-skipped related 1; ODDodd-skipped (Drosophila) homolog odd-skipped homolog odd-skipped related 1 (Drosophila) protein odd-skipped-related 1; Protein odd-skipped-related 1
Gene Aliases: ODD; OSR1
UniProt ID: (Human) Q8TAX0
Entrez Gene ID: (Human) 130497
Molecular Function:
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.