R&D Systems
Manufacturer Code:MAB4389
Catalog # MAB4389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 469221 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human OSM R beta Glu28-Ser739 Accession # Q99650 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IL-31 receptor subunit beta; IL-31 receptor subunit beta IL-31R subunit beta IL-31RB IL-31R-beta Interleukin-31 receptor subunit beta MGC150627 MGC75127 oncostatin M receptor oncostatin-M specific receptor beta subunit oncostatin-M-specific receptor subunit beta OSM R beta OSMR OSMRB OSMRBMGC150626 PLCA1; IL-31R subunit beta; Interleukin-31 receptor subunit beta; oncostatin-M specific receptor beta subunit; Oncostatin-M-specific receptor subunit beta
Gene Aliases: IL-31R-beta; IL-31RB; OSMR; OSMRB; PLCA1
UniProt ID: (Human) Q99650
Entrez Gene ID: (Human) 9180
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.