R&D Systems
Manufacturer Code:FAB662N100UG
Catalog # FAB662N100U
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 118125 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse OSM R beta Glu24-Leu738 Accession No. O70458 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Rat Monoclonal Antibody
Protein Aliases: IL-31 receptor subunit beta; IL-31 receptor subunit beta IL-31R subunit beta IL-31RB IL-31R-beta Interleukin-31 receptor subunit beta MGC150627 MGC75127 oncostatin M receptor oncostatin-M specific receptor beta subunit oncostatin-M-specific receptor subunit beta OSM R beta OSMR OSMRB OSMRBMGC150626 PLCA1; IL-31R subunit beta; Interleukin-31 receptor subunit beta; oncostatin-M specific receptor beta subunit; Oncostatin-M-specific receptor subunit beta
Gene Aliases: IL-31R-beta; IL-31RB; OSMR; OSMRB; PLCA1
UniProt ID: (Human) Q99650
Entrez Gene ID: (Human) 9180
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.