Invitrogen
Manufacturer Code:MA529458
Catalog # PIMA529458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8F3H8B2 |
Immunogen | Recombit Human OSMR protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IL-31 receptor subunit beta; IL-31R subunit beta; Interleukin-31 receptor subunit beta; oncostatin M receptor MGC150626 MGC150627 MGC75127; oncostatin-M specific receptor beta subunit; Oncostatin-M-specific receptor subunit beta
Gene Aliases: IL-31R-beta; IL-31RB; OSMR; OSMRB; PLCA1
UniProt ID: (Human) Q99650
Entrez Gene ID: (Human) 9180
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.