Invitrogen
Manufacturer Code:PA5117114
Catalog # PIPA5117114
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human OSGEPL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610001M19Rik AA416452 GCP1 HAMAP-Rule:MF_03179} N6-L-threonylcarbamoyladenine synthase OSGEPL1 O-sialoglycoprotein endopeptidase like 1 O-sialoglycoprotein endopeptidase-like 1 O-sialoglycoprotein endopeptidase-like protein 1 O-sialoglycoprotein endopeptidase-like protein 1 {ECO:0000255 probable O-sialoglycoprotein endopeptidase 2 probable tRNA N6-adenosine threonylcarbamoyltransferase mitochondrial probable tRNA N6-adenosine threonylcarbamoyltransferase mitochondrial {ECO:0000255 probable tRNA threonylcarbamoyladenosine biosynthesis protein OSGEPL1 putative sialoglycoprotease type 2 Qri7 t(6)A synthase t(6)A37 threonylcarbamoyladenosine biosynthesis protein Osgepl1 t(6)A37 threonylcarbamoyladenosine biosynthesis protein Osgepl1 {ECO:0000255 tRNA threonylcarbamoyladenosine biosynthesis protein Osgepl1 tRNA threonylcarbamoyladenosine biosynthesis protein Osgepl1 {ECO:0000255; N6-L-threonylcarbamoyladenine synthase; O-sialoglycoprotein endopeptidase like 1; O-sialoglycoprotein endopeptidase-like protein 1; O-sialoglycoprotein endopeptidase-like protein 1 {ECO:0000255|HAMAP-Rule:MF_03179}; OSGEPL1; probable O-sialoglycoprotein endopeptidase 2; Probable tRNA N6-adenosine threonylcarbamoyltransferase, mitochondrial; probable tRNA N6-adenosine threonylcarbamoyltransferase, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03179}; probable tRNA threonylcarbamoyladenosine biosynthesis protein OSGEPL1; putative sialoglycoprotease type 2; t(6)A synthase; t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEPL1; t(6)A37 threonylcarbamoyladenosine biosynthesis protein Osgepl1 {ECO:0000255|HAMAP-Rule:MF_03179}; tRNA threonylcarbamoyladenosine biosynthesis protein OSGEPL1; tRNA threonylcarbamoyladenosine biosynthesis protein Osgepl1 {ECO:0000255|HAMAP-Rule:MF_03179}
Gene Aliases: 2610001M19Rik; AA416452; GCP1; OSGEPL1; Qri7
UniProt ID: (Human) Q9H4B0, (Mouse) Q6PEB4, (Rat) Q4V7F3
Entrez Gene ID: (Human) 64172, (Mouse) 72085, (Rat) 314548
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.