Novus Biologicals
Manufacturer Code:NBP29375120UL
Catalog # NB029365
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human OSGEPL1 (NP_071748.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.24.57 O-sialoglycoprotein endopeptidase like 1 O-sialoglycoprotein endopeptidase-like 1 O-sialoglycoprotein endopeptidase-like protein 1 probable O-sialoglycoprotein endopeptidase 2 putative sialoglycoprotease type 2 t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEPL1; N6-L-threonylcarbamoyladenine synthase; O-sialoglycoprotein endopeptidase like 1; O-sialoglycoprotein endopeptidase-like protein 1; OSGEPL1; probable O-sialoglycoprotein endopeptidase 2; Probable tRNA N6-adenosine threonylcarbamoyltransferase, mitochondrial; probable tRNA threonylcarbamoyladenosine biosynthesis protein OSGEPL1; putative sialoglycoprotease type 2; t(6)A synthase; t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEPL1; tRNA threonylcarbamoyladenosine biosynthesis protein OSGEPL1
Gene Aliases: GCP1; OSGEPL1; Qri7
UniProt ID: (Human) Q9H4B0
Entrez Gene ID: (Human) 64172
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.