Proteintech
Manufacturer Code:256941AP150UL
Catalog # 50-173-5028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | OSGEPL1 Fusion Protein Ag22204 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: N6-L-threonylcarbamoyladenine synthase; O-sialoglycoprotein endopeptidase like 1; O-sialoglycoprotein endopeptidase-like protein 1; OSGEPL1; probable O-sialoglycoprotein endopeptidase 2; Probable tRNA N6-adenosine threonylcarbamoyltransferase, mitochondrial; probable tRNA threonylcarbamoyladenosine biosynthesis protein OSGEPL1; putative sialoglycoprotease type 2; t(6)A synthase; t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEPL1; tRNA threonylcarbamoyladenosine biosynthesis protein OSGEPL1
Gene Aliases: 2610001M19Rik; AA416452; GCP1; OSGEPL1; Qri7
UniProt ID: (Human) Q9H4B0, (Mouse) Q6PEB4
Entrez Gene ID: (Human) 64172, (Mouse) 72085
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.