Novus Biologicals
Manufacturer Code:NBP293164100UL
Catalog # NB028215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-335 of human OSGEP (NP_060277.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.24.57 GCPL1FLJ20411 hOSGEP KAE1 OSGEP1 O-sialoglycoprotein endopeptidase probable O-sialoglycoprotein endopeptidase PRSMG1 t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEP; hOSGEP; N6-L-threonylcarbamoyladenine synthase; O-sialoglycoprotein endopeptidase; OSGEP; probable O-sialoglycoprotein endopeptidase; Probable tRNA N6-adenosine threonylcarbamoyltransferase; probable tRNA threonylcarbamoyladenosine biosynthesis protein OSGEP; t(6)A synthase; t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEP; tRNA threonylcarbamoyladenosine biosynthesis protein OSGEP
Gene Aliases: GCPL1; KAE1; OSGEP; OSGEP1; PRSMG1
UniProt ID: (Human) Q9NPF4
Entrez Gene ID: (Human) 55644
Molecular Function: hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.