Novus Biologicals
Manufacturer Code:NBP238858
Catalog # NBP238858
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: LCGLPASATVHPDQDLFPLNGSSLENDAFSDKSERENPEESDTETQDHSRKTESGSDQSETPGAPVRRGTTYVEQVQEEL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.2.1 EC 2.7.7.6 FLJ42929 KIAA1534oxysterol-binding protein-related protein 5 OBPH1 ORP-5 ORP5oxysterol-binding protein homologue 1 OSBP-related protein 5 oxysterol binding protein-like 5 oxysterol-binding protein homolog 1 Oxysterol-binding protein homolog 1; ORP-5; OSBP-related protein 5; oxysterol binding protein-like 5; Oxysterol-binding protein homolog 1; oxysterol-binding protein homologue 1; Oxysterol-binding protein-related protein 5
Gene Aliases: KIAA1534; OBPH1; ORP5; OSBPL5
UniProt ID: (Human) Q9H0X9
Entrez Gene ID: (Human) 114879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.