Abnova
Manufacturer Code:PAB6516
Catalog # 89-132-466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human OSBPL5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of OSBPL5.
For Research Use Only
Protein Aliases: FLJ42929; OBPH1; ORP-5; ORP5; OSBP-related protein 5; oxysterol binding protein-like 5; Oxysterol-binding protein homolog 1; oxysterol-binding protein homologue 1; Oxysterol-binding protein-related protein 5
Gene Aliases: KIAA1534; OBPH1; ORP5; OSBPL5
UniProt ID: (Human) Q9H0X9
Entrez Gene ID: (Human) 114879
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.