Novus Biologicals
Manufacturer Code:NBP203327
Catalog # NBP203327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6A6 |
Immunogen | Full length human recombinant protein of human OSBPL11 (NP_073613) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ13012 FLJ13164 ORP11OSBP-related protein 11 ORP-11TCCCIA00292 OSBP12 oxysterol binding protein-like 11 oxysterol-binding protein-related protein 11; ORP-11; OSBP-related protein 11; oxysterol binding protein-like 11; Oxysterol-binding protein-related protein 11; testicular tissue protein Li 132
Gene Aliases: ORP-11; ORP11; OSBP12; OSBPL11; TCCCIA00292
UniProt ID: (Human) Q9BXB4
Entrez Gene ID: (Human) 114885
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.