Invitrogen
Manufacturer Code:PA530110
Catalog # PIPA530110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 154 and 428 of Human OSBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: OSBP1 oxysterol-binding protein 1; Oxysterol-binding protein 1
Gene Aliases: 1110018F06Rik; AW559088; Kiaa4220; mKIAA4220; OSBP; OSBP1
UniProt ID: (Human) P22059, (Mouse) Q3B7Z2
Entrez Gene ID: (Human) 5007, (Mouse) 76303
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.