Invitrogen
Manufacturer Code:MA534773
Catalog # PIMA534773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG38-07 |
Immunogen | Recombinant protein of C-terminal Human OS9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 4632413K17Rik Amplified in osteosarcoma 9 AU022351 ERLEC2 OS9 Os-9 Protein OS-9; Amplified in osteosarcoma 9; endoplasmic reticulum lectin 2; erlectin 2; osteosarcoma amplified 9, endoplasmic reticulum associated protein; osteosarcoma amplified 9, endoplasmic reticulum lectin; Protein OS-9
Gene Aliases: ERLEC2; OS-9; OS9
UniProt ID: (Human) Q13438
Entrez Gene ID: (Human) 10956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.