Invitrogen
Manufacturer Code:PA542628
Catalog # PIPA542628
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human OS9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amplified in osteosarcoma; Amplified in osteosarcoma 9; endoplasmic reticulum lectin 2; erlectin 2; osteosarcoma amplified 9; osteosarcoma amplified 9, endoplasmic reticulum associated protein; osteosarcoma amplified 9, endoplasmic reticulum lectin; Protein OS-9
Gene Aliases: 4632413K17Rik; AU022351; ERLEC2; OS-9; OS9
UniProt ID: (Human) Q13438, (Bovine) Q3MHX6, (Rat) Q5RKH6, (Mouse) Q8K2C7
Entrez Gene ID: (Human) 10956, (Rabbit) 100345796, (Horse) 100050776, (Dog) 474419, (Bovine) 504832, (Rat) 362891, (Guinea pig) 100718519, (Mouse) 216440
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.