Invitrogen
Manufacturer Code:PA522781
Catalog # PIPA522781
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of isoform 1 of the human OS9 protein (within residues 300-400). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amplified in osteosarcoma 9; endoplasmic reticulum lectin 2; ERLEC2 OS-9 amplified in osteosarcoma 9 endoplasmic reticulum lectin 2 erlectin 2 osteosarcoma amplified 9 endoplasmic reticulum associated protein protein OS-9; erlectin 2; osteosarcoma amplified 9; osteosarcoma amplified 9, endoplasmic reticulum associated protein; osteosarcoma amplified 9, endoplasmic reticulum lectin; Protein OS-9
Gene Aliases: 4632413K17Rik; AU022351; ERLEC2; OS-9; OS9
UniProt ID: (Human) Q13438, (Mouse) Q8K2C7
Entrez Gene ID: (Human) 10956, (Mouse) 216440
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.