Novus Biologicals
Manufacturer Code:NBP29396220UL
Catalog # NB022490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 353-612 of human OS9 (NP_001017956.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amplified in osteosarcoma 9; Amplified in osteosarcoma 9 endoplasmic reticulum lectin 2 ERLEC2 erlectin 2 OS-9 osteosarcoma amplified 9 endoplasmic reticulum associated protein osteosarcoma amplified 9 endoplasmic reticulum lectin protein OS-9; endoplasmic reticulum lectin 2; erlectin 2; osteosarcoma amplified 9, endoplasmic reticulum associated protein; osteosarcoma amplified 9, endoplasmic reticulum lectin; Protein OS-9
Gene Aliases: ERLEC2; OS-9; OS9
UniProt ID: (Human) Q13438
Entrez Gene ID: (Human) 10956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.