Invitrogen
Manufacturer Code:PA569271
Catalog # PIPA569271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human ORNT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D13S327 HHH ORC1 ORNT1 mitochondrial ornithine transporter 1 ornithine transporter 1 solute carrier family 25 member 15; Mitochondrial ornithine transporter 1; ornithine transporter 1; ornithine transporter) member 15; ornithine transporter, mitochondrial 1; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 15; Solute carrier family 25 member 15
Gene Aliases: Ab1-114; D13S327; D630044L02Rik; HHH; ORC1; ORNT1; SLC25A15; SP1855
UniProt ID: (Human) Q9Y619, (Rat) A0A0G2K309, (Mouse) Q9WVD5
Entrez Gene ID: (Bovine) 532560, (Sheep) 100147785, (Human) 10166, (Dog) 485993, (Rabbit) 100356921, (Rat) 306574, (Guinea pig) 100730028, (Mouse) 18408
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.