Invitrogen
Manufacturer Code:PA5113561
Catalog # PIPA5113561
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Mitochondrial ornithine transporter 1 protein (1-301AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ab1-114 BOS_12353 D13S327 D630044L02Rik HHH LOC100356921 Mitochondrial ornithine transporter 1 ORC1 ornithine transporter 1 ornithine transporter) member 15 ornithine transporter mitochondrial 1 ORNT1 SLC25A15 solute carrier family 25 (mitochondrial carrier ornithine transporter) member 15 solute carrier family 25 (mitochondrial carrier ornithine transporter) member 15 Solute carrier family 25 member 15 SP1855; Mitochondrial ornithine transporter 1; ornithine transporter 1; ornithine transporter, mitochondrial 1; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 15; Solute carrier family 25 member 15
Gene Aliases: D13S327; D630044L02Rik; HHH; ORC1; ORNT1; SLC25A15; SP1855
UniProt ID: (Human) Q9Y619, (Mouse) Q9WVD5
Entrez Gene ID: (Human) 10166, (Mouse) 18408
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.