Invitrogen
Manufacturer Code:PA5110383
Catalog # PIPA5110383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-301 of human SLC25A15 (NP_055067.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ab1-114 BOS_12353 D13S327 D630044L02Rik HHH Mitochondrial ornithine transporter 1 ORC1 ORNT1 Slc25a15 Solute carrier family 25 member 15 SP1855; Mitochondrial ornithine transporter 1; ornithine transporter 1; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 15; Solute carrier family 25 member 15
Gene Aliases: D13S327; HHH; ORC1; ORNT1; SLC25A15; SP1855
UniProt ID: (Human) Q9Y619
Entrez Gene ID: (Human) 10166
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.