Proteintech
Manufacturer Code:111991AP150UL
Catalog # 50-172-6962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ORM2 Fusion Protein Ag1667 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGP 2; AGP 2 AGP B AGP B&prime AGP2 Alpha 1 acid glycoprotein 2 OMD 2 ORM2 orosomucoid 2; alpha-1-acid glycoprotein 1; Alpha-1-acid glycoprotein 2; alpha-1-acid glycoprotein, type 2; bM340H1.5 (orosomucoid 2); OMD 2; Orosomucoid-2
Gene Aliases: Agp-2; AGP-B; AGP-B'; Agp1; AGP2; Orm-2; ORM2
UniProt ID: (Human) P19652, (Mouse) P07361
Entrez Gene ID: (Human) 5005, (Mouse) 18406
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.