Proteintech
Manufacturer Code:164391AP150UL
Catalog # 50-173-0943
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ORM1 Fusion Protein Ag9758 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGP 1; AGP 1 AGP A AGP1 Alpha 1 acid glycoprotein 1 OMD 1 ORM ORM1 orosomucoid 1; Alpha-1-acid glycoprotein 1; alpha-1-acid glycoprotein-2; bM340H1.2 (orosomucoid 1); epididymis secretory sperm binding protein Li 153w; OMD 1; Orosomucoid-1
Gene Aliases: Agp-1; Agp-2; AGP-A; AGP1; HEL-S-153w; ORM; Orm-1; ORM1
UniProt ID: (Human) P02763, (Mouse) Q60590
Entrez Gene ID: (Human) 5004, (Mouse) 18405
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.