Novus Biologicals
Manufacturer Code:NBP221065
Catalog # NBP221065
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic Peptide CKTSETVPRPA conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kappa-type 3 opioid receptor; Kappa-type 3 opioid receptor KOR-3MGC34578 nociceptin receptor NOCIR OORORL1kappa3-related opioid receptor opiate receptor-like 1 Orphanin FQ receptor; kappa3-related opioid receptor; KOR-3; Nociceptin receptor; nociceptin/orphanin FQ receptor; opiate receptor-like 1; Orphanin FQ receptor
Gene Aliases: KOR-3; NOCIR; NOP; NOPr; OOR; OPRL1; ORL1
UniProt ID: (Human) P41146
Entrez Gene ID: (Human) 4987
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.