Invitrogen
Manufacturer Code:PA528083
Catalog # PIPA528083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 648 and 711 of Human ORC3L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of latheo, Drosophila; LAT LATHEO ORC3L homolog of latheo Drosophila origin recognition complex subunit 3 origin recognition complex subunit Latheo origin recognition complex subunit 3 honolog origin recognition complex subunit 3-like; Origin recognition complex subunit 3; Origin recognition complex subunit Latheo; origin recognition complex, subunit 3 honolog
Gene Aliases: LAT; LATHEO; ORC3; ORC3L
UniProt ID: (Human) Q9UBD5
Entrez Gene ID: (Human) 23595
Molecular Function:
DNA binding protein
nucleic acid binding
replication origin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.