Novus Biologicals
Manufacturer Code:NB100121FR
Catalog # NB100121FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ORC1 7F6/1 |
Immunogen | Bacterially expressed Human ORC1 |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HS HSORC1 ORC1Lorigin recognition complex subunit 1 origin recognition complex subunit 1 origin recognition complex subunit 1 (yeast homolog)-like origin recognition complex subunit 1 homolog origin recognition complex subunit 1 homolog (S. cerevisiae) origin recognition complex subunit 1-like origin recognition complex subunit 1-like (yeast) PARC1origin recognition complex 1 Replication control protein 1; Origin recognition complex subunit 1; origin recognition complex, subunit 1 homolog; Replication control protein 1
Gene Aliases: HSORC1; ORC1; ORC1L; PARC1
UniProt ID: (Human) Q13415
Entrez Gene ID: (Human) 4998
Molecular Function:
DNA binding protein
nucleic acid binding
replication origin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.