Invitrogen
Manufacturer Code:PA575542
Catalog # PIPA575542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 364-409 of Human ORC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Origin recognition complex subunit 1; Origin recognition complex subunit 1 Replication control protein 1 ORC1L PARC1; origin recognition complex, subunit 1 homolog; origin recognition complex, subunit 1-like; Replication control protein 1
Gene Aliases: AA545195; HSORC1; MmORC1; ORC1; ORC1L; PARC1
UniProt ID: (Human) Q13415, (Rat) Q80Z32, (Mouse) Q9Z1N2
Entrez Gene ID: (Human) 4998, (Rat) 313479, (Mouse) 18392
Molecular Function:
DNA binding protein
nucleic acid binding
replication origin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.