Novus Biologicals
Manufacturer Code:NBP17998620UL
Catalog # NBP17998620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human OR13C9. Peptide sequence IFYGTILFMYMKPKSKETLNSDDLDATDKIISMFYGVMTPMMNPLIYSLR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Olfactory receptor 13C9; olfactory receptor family 13 subfamily C member 9 OR37L; Olfactory receptor OR9-13; olfactory receptor, family 13, subfamily C, member 9
Gene Aliases: OR13C9; OR37L; OR9-13
UniProt ID: (Human) Q8NGT0
Entrez Gene ID: (Human) 286362
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.