Invitrogen
Manufacturer Code:PA538164
Catalog # PIPA538164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human OR13C4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSHTPCRX17 HTPCRX17 OR2K1 OR37F OR9-7 olfactory receptor 13C4 olfactory receptor OR9-7 olfactory receptor family 2 subfamily K member 1; Olfactory receptor 13C4; Olfactory receptor OR9-7; olfactory receptor, family 13, subfamily C, member 4; olfactory receptor, family 2, subfamily K, member 1
Gene Aliases: HSHTPCRX17; HTPCRX17; OR13C4; OR2K1; OR37F; OR9-7
UniProt ID: (Human) Q8NGS5
Entrez Gene ID: (Human) 138804
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.